Assay Details
Target Gene Details
Entrez Gene ID: | 11057 |
Gene Name: | abhydrolase domain containing 2 |
Gene Aliases: |
HS1-2, LABH2, PHPS1-2 |
Location: |
Chr.15:89088150-89202360 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD2 | NM_007011.7 | NP_008942.3 | ||
| NM_152924.4 | NP_690888.1 | |||
| XM_005254832.1 | XP_005254889.1 | |||
| XM_005254833.1 | XP_005254890.1 | |||
| XM_011521165.1 | XP_011519467.1 | |||
| AF546700.1 | AAQ12021.1 | |||
| AK222841.1 | BAD96561.1 | |||
| AK223083.1 | BAD96803.1 | |||
| AK290340.1 | ||||
| AK296827.1 | ||||
| AL110291.1 | ||||
| BC019248.1 | AAH19248.1 | |||
| BC090052.1 | AAH90052.1 | |||
| BI464471.1 | ||||
| X12433.1 | CAA30976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv459e214 | Chr.15:89038688 - 89124261 on Build GRCh38 | Gain |
|
| nsv1050740 | Chr.15:89006295 - 89155865 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map