Assay Details
Target Gene Details
Entrez Gene ID: | 196951 |
Gene Name: | family with sequence similarity 227 member B |
Gene Aliases: |
C15orf33 |
Location: |
Chr.15:49326997-49620931 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | XM_005254279.3 | XP_005254336.1 | ||
| XM_005254280.3 | XP_005254337.1 | |||
| XM_005254284.3 | XP_005254341.1 | |||
| XM_006720462.3 | XP_006720525.1 | |||
| XM_006720463.3 | XP_006720526.1 | |||
| XM_006720464.3 | XP_006720527.1 | |||
| XM_011521441.2 | XP_011519743.1 | |||
| XM_017022063.1 | XP_016877552.1 | |||
| XM_017022064.1 | XP_016877553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833002 | Chr.15:49329203 - 49489602 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map