Assay Details
Target Gene Details
Entrez Gene ID: | 4916 |
Gene Name: | neurotrophic receptor tyrosine kinase 3 |
Gene Aliases: |
GP145-TrkC, TRKC, gp145(trkC) |
Location: |
Chr.15:87859749-88256796 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK3 | XM_006720543.3 | 17 | 12875 | XP_006720606.1 |
| XM_006720544.3 | 16 | 12851 | XP_006720607.1 | |
| XM_017022240.1 | 18 | 12746 | XP_016877729.1 | |
| XM_017022241.1 | 17 | 12231 | XP_016877730.1 | |
| XM_017022243.1 | 17 | 12227 | XP_016877732.1 | |
| XM_017022244.1 | 17 | 12129 | XP_016877733.1 | |
| XM_017022245.1 | 18 | 12184 | XP_016877734.1 | |
| XM_017022246.1 | 17 | 12285 | XP_016877735.1 | |
| XM_017022251.1 | 10 | 11246 | XP_016877740.1 | |
| XM_017022252.1 | 9 | 11222 | XP_016877741.1 | |
| XM_017022253.1 | 18 | 12917 | XP_016877742.1 | |
| XM_017022254.1 | 18 | 12950 | XP_016877743.1 |
More Information
Set Membership: |
|

Genomic Map