Assay Details
Target Gene Details
Entrez Gene ID: | 5432 |
Gene Name: | RNA polymerase II subunit C |
Gene Aliases: |
RPB3, RPB31, hRPB33, hsRPB3 |
Location: |
Chr.16:57462639-57472009 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2C | NM_032940.2 | NP_116558.1 | ||
| AF008443.1 | AAC14355.1 | |||
| AJ224143.1 | CAA11842.1 | |||
| AJ224144.1 | CAA11843.1 | |||
| AK293112.1 | ||||
| AK295582.1 | ||||
| BC000409.2 | AAH00409.1 | |||
| BC003159.1 | AAH03159.1 | |||
| BC028157.1 | AAH28157.1 | |||
| BI458170.1 | ||||
| CR542041.1 | CAG46838.1 | |||
| J05448.1 | AAA36586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map