Assay Details
Target Gene Details
Entrez Gene ID: | 4775 |
Gene Name: | nuclear factor of activated T-cells 3 |
Gene Aliases: |
NFAT4, NFATX |
Location: |
Chr.16:68085366-68229259 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFATC3 | NM_004555.3 | NP_004546.1 | ||
NM_173163.2 | NP_775186.1 | |||
NM_173165.2 | NP_775188.1 | |||
AB209824.1 | BAD93061.1 | |||
AK290370.1 | ||||
BC001050.2 | AAH01050.1 | |||
BU849493.1 | ||||
EU887605.1 | ||||
EU887606.1 | ||||
EU887607.1 | ||||
EU887608.1 | ||||
EU887609.1 | ||||
EU887610.1 | ||||
EU887611.1 | ||||
EU887612.1 | ||||
EU887613.1 | ||||
EU887614.1 | ||||
EU887615.1 | ||||
EU887616.1 | ||||
EU887617.1 | ||||
EU887618.1 | ||||
EU887619.1 | ||||
EU887620.1 | ||||
EU887621.1 | ||||
EU887622.1 | ||||
EU887623.1 | ||||
EU887624.1 | ||||
EU887625.1 | ||||
L41067.1 | AAA79174.1 | |||
U14510.1 | AAA86308.1 | |||
U85428.1 | AAB46595.1 | |||
U85429.1 | AAB46596.1 | |||
U85430.1 | AAB46597.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
