Assay Details
Target Gene Details
Entrez Gene ID: | 2903 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2A |
Gene Aliases: |
EPND, FESD, GluN2A, LKS, NMDAR2A, NR2A |
Location: |
Chr.16:9753404-10183364 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2A | NM_000833.4 | 14 | 9097 | NP_000824.1 |
| NM_001134407.2 | 13 | 9335 | NP_001127879.1 | |
| NM_001134408.2 | 14 | 8653 | NP_001127880.1 | |
| XM_011522458.2 | 12 | 8583 | XP_011520760.1 | |
| XM_011522461.2 | 15 | 9364 | XP_011520763.1 | |
| XM_017023172.1 | 12 | 9208 | XP_016878661.1 | |
| XM_017023173.1 | 13 | 8865 | XP_016878662.1 | |
| CR749843.1 | 1 | 1943 |
More Information
Set Membership: |
|

Genomic Map