Assay Details
Target Gene Details
Entrez Gene ID: | 83752 |
Gene Name: | lon peptidase 2, peroxisomal |
Gene Aliases: |
LONP, LONPL |
Location: |
Chr.16:48244167-48353979 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LONP2 | NM_001300948.1 | NP_001287877.1 | ||
NM_031490.3 | NP_113678.2 | |||
XM_017023755.1 | XP_016879244.1 | |||
XM_017023756.1 | XP_016879245.1 | |||
AJ548761.1 | CAD68987.1 | |||
AK027544.1 | ||||
AK074775.1 | BAC11201.1 | |||
AK127125.1 | ||||
AK294779.1 | ||||
AL834201.1 | CAD38889.1 | |||
BC093910.1 | AAH93910.1 | |||
BC093912.1 | AAH93912.1 | |||
BC143246.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
