Assay Details
Target Gene Details
Entrez Gene ID: | 93517 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 1 |
Gene Aliases: |
HSPC105 |
Location: |
Chr.16:81988855-82011488 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR42E1 | NM_145168.2 | NP_660151.2 | ||
| XM_005256257.3 | XP_005256314.1 | |||
| AK026740.1 | ||||
| AK315652.1 | ||||
| BC019670.1 | AAH19670.1 | |||
| BX482154.1 |
More Information
Set Membership: |
|

Genomic Map