Assay Details
Target Gene Details
Entrez Gene ID: | 85445 |
Gene Name: | contactin associated protein like 4 |
Gene Aliases: |
CASPR4 |
Location: |
Chr.16:76277217-76563610 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CNTNAP4 | NM_001322178.1 | NP_001309107.1 | ||
NM_001322179.1 | NP_001309108.1 | |||
NM_001322180.1 | NP_001309109.1 | |||
NM_001322181.1 | NP_001309110.1 | |||
NM_001322187.1 | NP_001309116.1 | |||
NM_001322188.1 | NP_001309117.1 | |||
NM_001322189.1 | NP_001309118.1 | |||
NM_001322190.1 | NP_001309119.1 | |||
NM_001322191.1 | NP_001309120.1 | |||
NM_033401.4 | NP_207837.2 | |||
NR_136210.1 | ||||
NR_136213.1 | ||||
NR_136216.1 | ||||
XM_017023805.1 | XP_016879294.1 | |||
AB051550.2 | ||||
AB100092.1 | BAC55270.1 | |||
AB100094.1 | BAC55272.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3892895 | Chr.16:76153094 - 76478682 on Build GRCh38 | Gain |
![]() |
nsv1065644 | Chr.16:76132681 - 76441737 on Build GRCh38 | Gain |
![]() |
nsv1064607 | Chr.16:75753231 - 76415204 on Build GRCh38 | Gain |
![]() |
nsv1059162 | Chr.16:75895450 - 76696919 on Build GRCh38 | Gain |
![]() ![]() |
dgv5192n54 | Chr.16:75901127 - 76701352 on Build GRCh38 | Gain |
![]() ![]() |
nsv833285 | Chr.16:76102447 - 76311501 on Build GRCh38 | Gain |
![]() |
esv2763142 | Chr.16:76237762 - 76427944 on Build GRCh38 | Loss |
![]() |
nsv833286 | Chr.16:76209832 - 76410386 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
