Assay Details
Target Gene Details
Entrez Gene ID: | 55715 |
Gene Name: | docking protein 4 |
Gene Aliases: |
IRS-5, IRS5 |
Location: |
Chr.16:57471958-57486473 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DOK4 | NM_018110.3 | NP_060580.2 | ||
XM_005256042.3 | XP_005256099.1 | |||
XM_005256043.2 | XP_005256100.1 | |||
AF466369.1 | AAL74195.1 | |||
AK001350.1 | BAA91642.1 | |||
AK307270.1 | ||||
AK315426.1 | ||||
BC001540.2 | AAH01540.1 | |||
BC003541.1 | AAH03541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
