Assay Details
Target Gene Details
Entrez Gene ID: | 57496 |
Gene Name: | MKL1/myocardin like 2 |
Gene Aliases: |
MRTF-B, MRTFB, NPD001 |
Location: |
Chr.16:14071076-14266779 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MKL2 | NM_001308142.1 | NP_001295071.1 | ||
| NM_014048.4 | NP_054767.3 | |||
| XM_005255452.3 | XP_005255509.1 | |||
| XM_006720909.3 | XP_006720972.1 | |||
| XM_006720913.2 | XP_006720976.1 | |||
| XM_006720914.2 | XP_006720977.1 | |||
| XM_011522568.2 | XP_011520870.1 | |||
| XM_017023501.1 | XP_016878990.1 | |||
| AK126948.1 | ||||
| AK294765.1 | ||||
| AY374297.1 | AAQ82435.1 | |||
| DB114294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571507 | Chr.16:14004283 - 14076712 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map