Assay Details
Target Gene Details
Entrez Gene ID: | 9100 |
Gene Name: | ubiquitin specific peptidase 10 |
Gene Aliases: |
UBPO |
Location: |
Chr.16:84699949-84779922 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP10 | NM_001272075.1 | NP_001259004.1 | ||
NM_005153.2 | NP_005144.2 | |||
NR_073577.1 | ||||
NR_073578.1 | ||||
XM_006721332.1 | XP_006721395.1 | |||
XM_011523440.1 | XP_011521742.1 | |||
XM_011523441.1 | XP_011521743.1 | |||
XM_011523443.1 | XP_011521745.1 | |||
XM_017023863.1 | XP_016879352.1 | |||
XM_017023864.1 | XP_016879353.1 | |||
XM_017023865.1 | XP_016879354.1 | |||
XM_017023866.1 | XP_016879355.1 | |||
XM_017023867.1 | XP_016879356.1 | |||
XM_017023868.1 | XP_016879357.1 | |||
XM_017023869.1 | XP_016879358.1 | |||
AK293304.1 | ||||
AK299618.1 | ||||
AK315570.1 | ||||
AL162049.1 | CAB82392.2 | |||
BC000263.1 | AAH00263.1 | |||
BX537402.1 | CAD97644.1 | |||
CR749515.1 | CAH18330.1 | |||
D80012.1 | ||||
DB077919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1061271 | Chr.16:84707543 - 84729856 on Build GRCh38 | Loss |
![]() |
nsv483044 | Chr.16:84166395 - 87066394 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
