Assay Details
Target Gene Details
Entrez Gene ID: | 55239 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 1 |
Gene Aliases: |
TPA1 |
Location: |
Chr.16:56451480-56477495 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGFOD1 | NM_001324357.1 | NP_001311286.1 | ||
| NM_001324358.1 | 1 | 285 | NP_001311287.1 | |
| NM_001324359.1 | NP_001311288.1 | |||
| NM_001324360.1 | NP_001311289.1 | |||
| NM_001324361.1 | NP_001311290.1 | |||
| NM_001324362.1 | NP_001311291.1 | |||
| NM_001324363.1 | NP_001311292.1 | |||
| NM_018233.3 | NP_060703.3 | |||
| AB046832.1 | ||||
| AK001688.1 | BAA91838.1 | |||
| AK022130.1 | BAB13967.1 | |||
| AK022752.1 | BAB14226.1 | |||
| AK304584.1 | ||||
| AL711389.1 | ||||
| BC032919.1 | AAH32919.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map