Assay Details
Target Gene Details
Entrez Gene ID: | 29070 |
Gene Name: | coiled-coil domain containing 113 |
Gene Aliases: |
HSPC065 |
Location: |
Chr.16:58196863-58283836 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC113 | XM_011523039.2 | XP_011521341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv572763 | Chr.16:58173299 - 58234532 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map