Assay Details
Target Gene Details
Entrez Gene ID: | 8448 |
Gene Name: | double C2 domain alpha |
Gene Aliases: |
Doc2 |
Location: |
Chr.16:30005514-30023280 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOC2A | XM_011545975.1 | 1 | 229 | XP_011544277.1 |
| XM_017023776.1 | 1 | 229 | XP_016879265.1 |
More Information
Set Membership: |
|

Genomic Map