Assay Details
Target Gene Details
Entrez Gene ID: | 115584 |
Gene Name: | solute carrier family 5 member 11 |
Gene Aliases: |
KST1, RKST1, SGLT6, SMIT2 |
Location: |
Chr.16:24832784-24911711 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC5A11 | XM_017022902.1 | XP_016878391.1 |
More Information
Set Membership: |
|

Genomic Map