Assay Details
Target Gene Details
Entrez Gene ID: | 101926950 |
Gene Name: | long intergenic non-protein coding RNA 1570 |
Gene Aliases: |
- |
Location: |
Chr.16:5601169-5616250 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC01570 | NR_110902.1 | |||
| DB449025.1 |
Target Gene Details
Entrez Gene ID: | 54715 |
Gene Name: | RNA binding protein, fox-1 homolog 1 |
Gene Aliases: |
2BP1, A2BP1, FOX-1, FOX1, HRNBP1 |
Location: |
Chr.16:5239749-7713343 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBFOX1 | XM_017023318.1 | XP_016878807.1 | ||
| XM_017023319.1 | XP_016878808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035156 | Chr.16:5091032 - 5996934 on Build GRCh38 | Gain |
|
| nsv571308 | Chr.16:5535851 - 5969940 on Build GRCh38 | Gain |
|
| nsv510675 | Chr.16:5543389 - 5609986 on Build GRCh38 | Deletion |
|
| nsv833133 | Chr.16:5533052 - 5681669 on Build GRCh38 | Loss |
|
| nsv1048972 | Chr.16:5466061 - 5752374 on Build GRCh38 | Gain |
|
| nsv1042301 | Chr.16:5531685 - 5969222 on Build GRCh38 | Gain |
|
| esv3892765 | Chr.16:5529214 - 5699442 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map