Assay Details
Target Gene Details
Entrez Gene ID: | 729993 |
Gene Name: | shisa family member 9 |
Gene Aliases: |
CKAMP44 |
Location: |
Chr.16:12901619-13563427 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHISA9 | NM_001145204.2 | 1 | 48 | NP_001138676.2 |
| NM_001145205.1 | 1 | 48 | NP_001138677.2 | |
| XM_005255539.3 | 1 | 49 | XP_005255596.2 | |
| XM_011522642.2 | 1 | 49 | XP_011520944.1 | |
| AK299610.1 | ||||
| DC375222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833142 | Chr.16:12828715 - 13005564 on Build GRCh38 | Loss |
|
| nsv827557 | Chr.16:12900164 - 12903906 on Build GRCh38 | Gain |
|
| nsv952925 | Chr.16:12898644 - 12904843 on Build GRCh38 | Deletion |
|
| nsv1070329 | Chr.16:12901242 - 12901943 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map