Assay Details
Target Gene Details
Entrez Gene ID: | 54947 |
Gene Name: | lysophosphatidylcholine acyltransferase 2 |
Gene Aliases: |
AGPAT11, AYTL1, LysoPAFAT |
Location: |
Chr.16:55509001-55586670 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPCAT2 | NM_017839.4 | NP_060309.2 | ||
| XM_011523169.2 | XP_011521471.1 | |||
| XM_017023376.1 | XP_016878865.1 | |||
| AB244718.1 | ||||
| AK000488.1 | BAA91199.1 | |||
| AK308386.1 | ||||
| BC002472.2 | AAH02472.2 | |||
| BX641069.1 | CAE46034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521337 | Chr.16:55510637 - 55585662 on Build GRCh38 | Loss |
|
| nsv1062908 | Chr.16:55514410 - 55569276 on Build GRCh38 | Loss |
|
| dgv2982n100 | Chr.16:55507884 - 55555666 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map