Assay Details
Target Gene Details
Entrez Gene ID: | 55512 |
Gene Name: | sphingomyelin phosphodiesterase 3 |
Gene Aliases: |
NSMASE2 |
Location: |
Chr.16:68358327-68448703 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPD3 | NM_018667.3 | 9 | 3358 | NP_061137.1 |
| XM_005256031.3 | 9 | 3451 | XP_005256088.1 | |
| XM_005256032.3 | 9 | 3470 | XP_005256089.1 | |
| XM_011523207.1 | 10 | 3451 | XP_011521509.1 | |
| XM_011523208.2 | 8 | 3552 | XP_011521510.1 | |
| XM_011523209.2 | 8 | 3335 | XP_011521511.1 | |
| XM_017023405.1 | 9 | 3277 | XP_016878894.1 | |
| XM_017023406.1 | 10 | 3758 | XP_016878895.1 | |
| XM_017023407.1 | 10 | 3480 | XP_016878896.1 | |
| XM_017023408.1 | 10 | 3434 | XP_016878897.1 | |
| AL157660.1 | 1 | 322 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582280 | Chr.16:68348088 - 68360930 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74024630] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map