Assay Details
Target Gene Details
Entrez Gene ID: | 113 |
Gene Name: | adenylate cyclase 7 |
Gene Aliases: |
AC7 |
Location: |
Chr.16:50246158-50318135 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY7 | XM_011522837.2 | XP_011521139.1 |
More Information
Set Membership: |
|

Genomic Map