Assay Details
Target Gene Details
Entrez Gene ID: | 23659 |
Gene Name: | phospholipase A2 group XV |
Gene Aliases: |
ACS, GXVPLA2, LLPL, LPLA2, LYPLA3 |
Location: |
Chr.16:68245344-68261062 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G15 | NM_012320.3 | NP_036452.1 | ||
| XM_005255886.3 | XP_005255943.1 | |||
| XM_011522979.2 | XP_011521281.1 | |||
| XM_011522980.2 | XP_011521282.2 | |||
| AB017494.1 | BAA76877.1 | |||
| AK001705.1 | ||||
| AK074828.1 | BAC11233.1 | |||
| AK127898.1 | ||||
| AK222790.1 | BAD96510.1 | |||
| AK296263.1 | ||||
| AK298494.1 | ||||
| AK300596.1 | ||||
| AL110209.1 | CAB53675.1 | |||
| AL389957.1 | CAB97531.1 | |||
| AY358425.1 | AAQ88791.1 | |||
| BC011640.2 | AAH11640.2 | |||
| BC062605.1 | AAH62605.1 |
More Information
Set Membership: |
|

Genomic Map