Assay Details
Target Gene Details
Entrez Gene ID: | 83752 |
Gene Name: | lon peptidase 2, peroxisomal |
Gene Aliases: |
LONP, LONPL |
Location: |
Chr.16:48244167-48353979 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LONP2 | NM_001300948.1 | NP_001287877.1 | ||
| NM_031490.3 | NP_113678.2 | |||
| XM_017023755.1 | XP_016879244.1 | |||
| XM_017023756.1 | XP_016879245.1 | |||
| AJ548761.1 | CAD68987.1 | |||
| AK027544.1 | ||||
| AK027666.1 | BAB55278.1 | |||
| AK074775.1 | BAC11201.1 | |||
| AK127125.1 | ||||
| AK294779.1 | ||||
| AL834201.1 | CAD38889.1 | |||
| BC093910.1 | AAH93910.1 | |||
| BC093912.1 | AAH93912.1 | |||
| BC110434.1 | ||||
| BC143246.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833219 | Chr.16:48256684 - 48425390 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map