Assay Details
Target Gene Details
Entrez Gene ID: | 92017 |
Gene Name: | sorting nexin 29 |
Gene Aliases: |
A-388D4.1, RUNDC2A |
Location: |
Chr.16:11976734-12574289 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX29 | NM_032167.4 | NP_115543.3 | ||
| XM_017023872.1 | XP_016879361.1 | |||
| XM_017023874.1 | XP_016879363.1 | |||
| XM_017023877.1 | XP_016879366.1 | |||
| AK022425.1 | BAB14033.1 | |||
| AK097928.1 | 1 | 1026 | ||
| AL135642.1 | ||||
| HY078299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043959 | Chr.16:11945489 - 12191412 on Build GRCh38 | Gain |
|
| nsv571458 | Chr.16:11831151 - 11993425 on Build GRCh38 | Gain |
|
| nsv527197 | Chr.16:11977484 - 11979943 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map