Assay Details
Target Gene Details
Entrez Gene ID: | 113 |
Gene Name: | adenylate cyclase 7 |
Gene Aliases: |
AC7 |
Location: |
Chr.16:50246158-50318135 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY7 | NM_001286057.1 | NP_001272986.1 | ||
| XM_011522836.2 | XP_011521138.1 | |||
| XM_011522837.2 | XP_011521139.1 | |||
| XM_011522840.2 | XP_011521142.1 | |||
| XM_011522842.2 | XP_011521144.1 | |||
| XM_017022895.1 | XP_016878384.1 | |||
| XM_017022897.1 | XP_016878386.1 | |||
| XM_017022898.1 | XP_016878387.1 | |||
| XM_017022899.1 | XP_016878388.1 | |||
| AK093706.1 | ||||
| BP270410.1 | ||||
| DA047753.1 |
More Information
Set Membership: |
|

Genomic Map