Assay Details
Target Gene Details
Entrez Gene ID: | 583 |
Gene Name: | Bardet-Biedl syndrome 2 |
Gene Aliases: |
BBS, RP74 |
Location: |
Chr.16:56470403-56520096 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BBS2 | XM_005256080.2 | XP_005256137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833243 | Chr.16:56375371 - 56549461 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map