Assay Details
Target Gene Details
Entrez Gene ID: | 1236 |
Gene Name: | C-C motif chemokine receptor 7 |
Gene Aliases: |
BLR2, CC-CKR-7, CCR-7, CD197, CDw197, CMKBR7, EBI1 |
Location: |
Chr.17:40553769-40565484 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR7 | NM_001301714.1 | NP_001288643.1 | ||
| NM_001301716.1 | NP_001288645.1 | |||
| NM_001301717.1 | NP_001288646.1 | |||
| NM_001301718.1 | NP_001288647.1 | |||
| NM_001838.3 | NP_001829.1 | |||
| AK127810.1 | ||||
| AK310277.1 | ||||
| AK312732.1 | ||||
| AY587876.1 | AAT52232.1 | |||
| BC035343.1 | AAH35343.1 | |||
| BI910219.1 | ||||
| DA814913.1 | ||||
| HY017993.1 | ||||
| HY021417.1 | ||||
| L08176.1 | AAA58615.1 | |||
| L31581.1 | AAA74231.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76372077] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map