Assay Details
Target Gene Details
Entrez Gene ID: | 9098 |
Gene Name: | ubiquitin specific peptidase 6 |
Gene Aliases: |
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short |
Location: |
Chr.17:5116438-5175032 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP6 | NM_001304284.1 | NP_001291213.1 | ||
| BX647719.1 |
Target Gene Details
Entrez Gene ID: | 7775 |
Gene Name: | zinc finger protein 232 |
Gene Aliases: |
ZSCAN11 |
Location: |
Chr.17:5105734-5123116 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF232 | NM_001320953.1 | NP_001307882.1 | ||
| XM_017025021.1 | XP_016880510.1 | |||
| AK316369.1 | ||||
| BQ641476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map