Assay Details
Target Gene Details
Entrez Gene ID: | 9482 |
Gene Name: | syntaxin 8 |
Gene Aliases: |
CARB |
Location: |
Chr.17:9250471-9575958 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STX8 | NM_004853.2 | NP_004844.1 | ||
NR_033656.1 | ||||
XM_011524079.2 | XP_011522381.1 | |||
AF036715.1 | AAC95285.1 | |||
AF062077.1 | AAC36466.1 | |||
AF115323.1 | AAD20831.1 | |||
BC009713.1 | AAH09713.1 | |||
BT007319.1 | AAP35983.1 | |||
BU596252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1064011 | Chr.17:9203289 - 9492596 on Build GRCh38 | Gain |
![]() ![]() |
nsv574341 | Chr.17:9148000 - 9504608 on Build GRCh38 | Gain |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
