Assay Details
Target Gene Details
Entrez Gene ID: | 9123 |
Gene Name: | solute carrier family 16 member 3 |
Gene Aliases: |
MCT 3, MCT 4, MCT-3, MCT-4, MCT3, MCT4 |
Location: |
Chr.17:82228406-82239499 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A3 | NM_001042422.2 | NP_001035887.1 | ||
| NM_001206950.1 | NP_001193879.1 | |||
| NM_004207.3 | NP_004198.1 | |||
| AK223040.1 | BAD96760.1 | |||
| BM926587.1 | ||||
| DA419716.1 | ||||
| DA583988.1 | ||||
| U81800.1 | AAC52015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2166 | Chr.17:82212292 - 82238972 on Build GRCh38 | Insertion |
|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv576193 | Chr.17:82226048 - 82237706 on Build GRCh38 | Loss |
|
| nsv833572 | Chr.17:82204977 - 82388835 on Build GRCh38 | Loss |
|
| esv21913 | Chr.17:81828123 - 83103548 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116990655] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map