Assay Details
Target Gene Details
Entrez Gene ID: | 101101775 |
Gene Name: | TMEM220 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:10729777-10815164 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM220-AS1 | NR_073455.1 | |||
| NR_073456.1 | ||||
| NR_073457.1 | ||||
| NR_073458.1 | ||||
| BQ694074.1 | ||||
| BQ694934.1 | ||||
| DA035282.1 | ||||
| DA190285.1 | ||||
| DA747858.1 | ||||
| DA867863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|

Genomic Map