Assay Details
Target Gene Details
Entrez Gene ID: | 708 |
Gene Name: | complement C1q binding protein |
Gene Aliases: |
GC1QBP, HABP1, SF2p32, gC1Q-R, gC1qR, p32 |
Location: |
Chr.17:5432779-5439151 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QBP | NM_001212.3 | NP_001203.1 | ||
| AF275902.1 | AAF78763.1 | |||
| AK291516.1 | ||||
| BC000435.2 | AAH00435.1 | |||
| BC013731.1 | AAH13731.1 | |||
| BT019898.1 | AAV38701.1 | |||
| BT019899.1 | AAV38702.1 | |||
| L04636.1 | AAA16315.1 | |||
| X75913.1 | CAA53512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map