Assay Details
Target Gene Details
Entrez Gene ID: | 9527 |
Gene Name: | golgi SNAP receptor complex member 1 |
Gene Aliases: |
GOLIM2, GOS-28, GOS28, GOS28/P28, GS28, P28 |
Location: |
Chr.17:30477402-30528314 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOSR1 | NM_001007024.1 | NP_001007025.1 | ||
| NM_001007025.1 | NP_001007026.1 | |||
| NM_004871.2 | NP_004862.1 | |||
| XM_005258070.4 | XP_005258127.1 | |||
| XM_005258071.4 | XP_005258128.1 | |||
| XM_005258072.2 | XP_005258129.1 | |||
| XM_017025374.1 | XP_016880863.1 | |||
| XM_017025375.1 | XP_016880864.1 | |||
| XM_017025376.1 | XP_016880865.1 | |||
| AF047438.1 | AAC39889.1 | |||
| AF073926.1 | AAD12945.1 | |||
| AK000606.1 | ||||
| AK291381.1 | ||||
| AK298714.1 | ||||
| BC040471.1 | AAH40471.1 | |||
| BG654545.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064611 | Chr.17:30466299 - 30587613 on Build GRCh38 | Gain |
|
| nsv515870 | Chr.17:30432017 - 30673001 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145839775] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map