Assay Details
Target Gene Details
Entrez Gene ID: | 3927 |
Gene Name: | LIM and SH3 protein 1 |
Gene Aliases: |
Lasp-1, MLN50 |
Location: |
Chr.17:38869859-38921770 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LASP1 | NM_001271608.1 | NP_001258537.1 | ||
| NM_006148.3 | NP_006139.1 | |||
| NR_073384.1 | ||||
| AK289847.1 | ||||
| AK294704.1 | ||||
| AK295522.1 | ||||
| AK296092.1 | ||||
| AK307868.1 | ||||
| AK309933.1 | ||||
| BC007560.1 | AAH07560.1 | |||
| BC012460.1 | AAH12460.1 | |||
| CN371189.1 | ||||
| DC324561.1 | ||||
| DC333179.1 | ||||
| X82456.1 | CAA57833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833439 | Chr.17:38743861 - 38923753 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs80253064] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map