Assay Details
Target Gene Details
Entrez Gene ID: | 100506215 |
Gene Name: | PAXBP1 antisense RNA 1 |
Gene Aliases: |
GCFC1-AS1 |
Location: |
Chr.21:32728114-32743122 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAXBP1-AS1 | NR_038879.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646891 | Chr.21:32675352 - 32751209 on Build GRCh38 | Gain |
|
| nsv834086 | Chr.21:32710449 - 32883526 on Build GRCh38 | Loss |
|

Genomic Map