Assay Details
Target Gene Details
Entrez Gene ID: | 10950 |
Gene Name: | BTG anti-proliferation factor 3 |
Gene Aliases: |
ANA, APRO4, TOB5, TOB55, TOFA |
Location: |
Chr.21:17593650-17612950 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTG3 | NM_001130914.1 | NP_001124386.1 | ||
| NM_006806.4 | NP_006797.3 | |||
| XM_011529441.1 | XP_011527743.1 | |||
| XM_017028265.1 | XP_016883754.1 | |||
| XM_017028266.1 | XP_016883755.1 | |||
| AK289408.1 | ||||
| BC011957.1 | AAH11957.1 | |||
| BT007276.1 | AAP35940.1 | |||
| CR457061.1 | CAG33342.1 | |||
| D64110.1 | BAA33788.1 | |||
| DR155504.1 |
Target Gene Details
Entrez Gene ID: | 1525 |
Gene Name: | coxsackie virus and adenovirus receptor |
Gene Aliases: |
CAR, CAR4/6, HCAR |
Location: |
Chr.21:17512906-17636213 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXADR | XM_011529476.2 | XP_011527778.1 | ||
| XM_011529477.2 | 6 | 3091 | XP_011527779.1 | |
| XM_011529478.2 | XP_011527780.1 | |||
| XM_011529479.1 | XP_011527781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575381 | Chr.21:17436662 - 17709158 on Build GRCh38 | Gain |
|
| esv3893382 | Chr.21:17364904 - 17698240 on Build GRCh38 | Gain |
|
| esv2723192 | Chr.21:16398054 - 17697109 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143649862] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map