Assay Details
Target Gene Details
Entrez Gene ID: | 1826 |
Gene Name: | DS cell adhesion molecule |
Gene Aliases: |
CHD2, CHD2-42, CHD2-52 |
Location: |
Chr.21:40010999-40847113 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAM | NM_001271534.1 | NP_001258463.1 | ||
| NM_001389.3 | NP_001380.2 | |||
| NR_073202.1 | ||||
| AB209136.1 | BAD92373.1 | |||
| AF023449.1 | AAC17966.1 | |||
| AF023450.1 | AAC17967.1 | |||
| AF217525.1 | AAF27525.1 |
Target Gene Details
Entrez Gene ID: | 100874326 |
Gene Name: | DSCAM intronic transcript 1 |
Gene Aliases: |
zag4 |
Location: |
Chr.21:40618507-40630767 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSCAM-IT1 | NR_046774.2 |
More Information
Set Membership: |
|

Genomic Map