Assay Details
Target Gene Details
Entrez Gene ID: | 10317 |
Gene Name: | beta-1,3-galactosyltransferase 5 |
Gene Aliases: |
B3GalT-V, B3GalTx, B3T5, GLCT5, beta-1,3-GalTase 5, beta-3-Gx-T5, beta3Gal-T5 |
Location: |
Chr.21:39612940-39662889 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALT5 | NM_001278650.1 | NP_001265579.1 | ||
| XM_017028235.1 | XP_016883724.1 | |||
| XM_017028236.1 | XP_016883725.1 | |||
| XM_017028237.1 | XP_016883726.1 | |||
| AK292951.1 | ||||
| AY372061.1 | AAR08910.1 | |||
| DQ645732.1 | ||||
| DQ645733.1 | ||||
| DQ645734.1 | ||||
| DQ645735.1 | ||||
| DQ645736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647023 | Chr.21:39622244 - 39639254 on Build GRCh38 | Loss |
|
| esv3647022 | Chr.21:39615180 - 39640459 on Build GRCh38 | Loss |
|
| nsv587485 | Chr.21:39628426 - 39656270 on Build GRCh38 | Loss |
|
| nsv1061454 | Chr.21:39386047 - 39893479 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs532990127] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map