Assay Details
Target Gene Details
Entrez Gene ID: | 757 |
Gene Name: | transmembrane protein 50B |
Gene Aliases: |
C21orf4, HCVP7TP3 |
Location: |
Chr.21:33432486-33480009 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM50B | NR_040016.1 | |||
| XM_011529746.1 | XP_011528048.1 | |||
| CB306801.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893406 | Chr.21:33437386 - 33486199 on Build GRCh38 | Loss |
|
| nsv834087 | Chr.21:33325890 - 33469002 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map