Assay Details
Target Gene Details
Entrez Gene ID: | 5435 |
Gene Name: | RNA polymerase II subunit F |
Gene Aliases: |
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15 |
Location: |
Chr.22:37953663-38041915 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR2F | NM_001301129.1 | NP_001288058.1 | ||
NM_001301130.1 | NP_001288059.1 | |||
NM_001301131.1 | NP_001288060.1 | |||
NM_021974.4 | NP_068809.1 | |||
NR_125371.1 | ||||
XM_017028832.1 | XP_016884321.1 | |||
AK094034.1 | ||||
AK311128.1 | ||||
AK311831.1 | ||||
BC003582.1 | AAH03582.1 | |||
BP195499.1 | ||||
CR456546.1 | CAG30432.1 | |||
CR457021.1 | CAG33302.1 | |||
DA083933.1 | ||||
DA203006.1 | ||||
DA372532.1 | ||||
DN989996.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
