Assay Details
Target Gene Details
Entrez Gene ID: | 23481 |
Gene Name: | pescadillo ribosomal biogenesis factor 1 |
Gene Aliases: |
PES |
Location: |
Chr.22:30576625-30607013 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PES1 | NM_001243225.1 | NP_001230154.1 | ||
| NM_001282327.1 | NP_001269256.1 | |||
| NM_001282328.1 | NP_001269257.1 | |||
| NM_014303.3 | NP_055118.1 | |||
| XM_006724173.1 | XP_006724236.1 | |||
| XM_017028678.1 | XP_016884167.1 | |||
| AB451380.1 | ||||
| AK096320.1 | ||||
| AK125778.1 | BAC86285.1 | |||
| AK127185.1 | ||||
| AK315518.1 | ||||
| BC032489.1 | AAH32489.1 | |||
| BQ576244.1 | ||||
| CR456539.1 | CAG30425.1 | |||
| U78310.1 | AAB61140.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588887 | Chr.22:30557534 - 30599369 on Build GRCh38 | Loss |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map