Assay Details
Target Gene Details
Entrez Gene ID: | 27443 |
Gene Name: | CECR2, histone acetyl-lysine reader |
Gene Aliases: |
- |
Location: |
Chr.22:17359949-17558155 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CECR2 | NM_001290046.1 | NP_001276975.1 | ||
| NM_001290047.1 | NP_001276976.1 | |||
| XM_011546128.2 | XP_011544430.1 | |||
| XM_011546129.2 | XP_011544431.1 | |||
| XM_011546132.2 | XP_011544434.1 | |||
| XM_017028785.1 | XP_016884274.1 | |||
| AF336133.1 | AAK15343.1 | |||
| AK308330.1 | ||||
| AL832377.1 | CAH56212.1 | |||
| BX647449.1 | CAH56122.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588099 | Chr.22:17333921 - 17436386 on Build GRCh38 | Gain |
|
| nsv834118 | Chr.22:17358660 - 17523783 on Build GRCh38 | Loss |
|
| esv3893427 | Chr.22:17214813 - 17405801 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map