Assay Details
Target Gene Details
Entrez Gene ID: | 25817 |
Gene Name: | family with sequence similarity 19 member A5, C-C motif chemokine like |
Gene Aliases: |
QLLK5208, TAFA-5, TAFA5, UNQ5208 |
Location: |
Chr.22:48489460-48751935 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A5 | NM_001082967.2 | NP_001076436.1 | ||
| AF131851.1 | AAD20061.1 | |||
| BC039396.1 | AAH39396.2 | |||
| DB481230.1 |
Target Gene Details
Entrez Gene ID: | 284933 |
Gene Name: | uncharacterized LOC284933 |
Gene Aliases: |
- |
Location: |
Chr.22:48538900-48547387 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC284933 | NR_038917.1 | 4 | 825 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv778n67 | Chr.22:48453824 - 48657701 on Build GRCh38 | Gain |
|
| nsv955186 | Chr.22:48484289 - 48570188 on Build GRCh38 | Deletion |
|
| nsv1064218 | Chr.22:48472061 - 48792098 on Build GRCh38 | Gain |
|
| nsv834232 | Chr.22:48358817 - 48553110 on Build GRCh38 | Loss |
|
| nsv589147 | Chr.22:48310704 - 48549198 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78011702] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map