Assay Details
Target Gene Details
Entrez Gene ID: | 5435 |
Gene Name: | RNA polymerase II subunit F |
Gene Aliases: |
HRBP14.4, POLRF, RPABC14.4, RPABC2, RPB14.4, RPB6, RPC15 |
Location: |
Chr.22:37953663-38041915 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2F | NM_001301129.1 | NP_001288058.1 | ||
| NM_001301130.1 | NP_001288059.1 | |||
| NM_001301131.1 | NP_001288060.1 | |||
| NM_021974.4 | NP_068809.1 | |||
| NR_125371.1 | ||||
| XM_017028832.1 | XP_016884321.1 | |||
| AK094034.1 | ||||
| AK311128.1 | ||||
| AK311831.1 | ||||
| BC003582.1 | AAH03582.1 | |||
| BP195499.1 | ||||
| CR456546.1 | CAG30432.1 | |||
| CR457021.1 | CAG33302.1 | |||
| DA083933.1 | ||||
| DA203006.1 | ||||
| DA372532.1 | ||||
| DN989996.1 |
More Information
Set Membership: |
|

Genomic Map