Assay Details
Target Gene Details
Entrez Gene ID: | 150280 |
Gene Name: | HORMA domain containing 2 |
Gene Aliases: |
CT46.2 |
Location: |
Chr.22:30080069-30207425 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HORMAD2 | XM_011529914.2 | XP_011528216.1 | ||
| XM_011529917.2 | XP_011528219.1 | |||
| XM_011529918.1 | XP_011528220.1 | |||
| XM_017028621.1 | XP_016884110.1 | |||
| XM_017028622.1 | XP_016884111.1 | |||
| XM_017028624.1 | XP_016884113.1 | |||
| AK098703.1 | BAC05388.1 | |||
| BC030013.1 | AAH30013.1 | |||
| BI459310.1 | ||||
| CR456341.1 | CAG30227.1 | |||
| HY002382.1 |
More Information
Set Membership: |
|

Genomic Map