Assay Details
Target Gene Details
Entrez Gene ID: | 340595 |
Gene Name: | zinc finger CCHC-type containing 16 |
Gene Aliases: |
Mar4, Mart4 |
Location: |
Chr.X:112083025-112457245 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZCCHC16 | NM_001004308.2 | NP_001004308.2 | ||
| AK128465.1 | BAC87453.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs139401874] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map