Assay Details
Target Gene Details
Entrez Gene ID: | 158763 |
Gene Name: | Rho GTPase activating protein 36 |
Gene Aliases: |
- |
Location: |
Chr.X:131058242-131089885 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP36 | NM_001282607.1 | NP_001269536.1 | ||
| NM_144967.3 | NP_659404.2 | |||
| AK128203.1 | BAC87322.1 | |||
| AK296748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523130 | Chr.X:130937509 - 131062354 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map