Assay Details
Target Gene Details
Entrez Gene ID: | 1730 |
Gene Name: | diaphanous related formin 2 |
Gene Aliases: |
DIA, DIA2, DRF2, POF, POF2 |
Location: |
Chr.X:96684663-97600598 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH2 | NM_006729.4 | NP_006720.1 | ||
| NM_007309.3 | NP_009293.1 | |||
| AK291272.1 | ||||
| BC117414.1 | ||||
| BC143838.1 | ||||
| BX363708.2 | ||||
| Y15908.1 | CAA75869.1 | |||
| Y15909.1 | CAA75870.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73250716] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map