Assay Details
Target Gene Details
Entrez Gene ID: | 7224 |
Gene Name: | transient receptor potential cation channel subfamily C member 5 |
Gene Aliases: |
PPP1R159, TRP5 |
Location: |
Chr.X:111774314-112082943 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC5 | NM_012471.2 | NP_036603.1 | ||
| XM_017029774.1 | XP_016885263.1 | |||
| AB209258.1 | BAD92495.1 | |||
| AF054568.1 | AAF00002.1 | |||
| BC137271.1 | ||||
| BC137274.1 |
Target Gene Details
Entrez Gene ID: | 100329135 |
Gene Name: | TRPC5 opposite strand |
Gene Aliases: |
TRPC5-AS1 |
Location: |
Chr.X:111876051-111903990 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC5OS | NM_001195576.1 | 4 | 2216 | NP_001182505.1 |
| NM_001195577.1 | 4 | 2197 | NP_001182506.1 | |
| NM_001195578.1 | 4 | 2151 | NP_001182507.1 | |
| XM_011530838.2 | 4 | 2145 | XP_011529140.1 | |
| BC034811.1 | 4 | 2197 |
More Information
Set Membership: |
|

Genomic Map