Assay Details
Target Gene Details
Entrez Gene ID: | 340547 |
Gene Name: | V-set and immunoglobulin domain containing 1 |
Gene Aliases: |
1700062D20Rik, GPA34, dJ889N15.1 |
Location: |
Chr.X:108018886-108079184 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG1 | XM_011530936.2 | XP_011529238.1 |
More Information
Set Membership: |
|

Genomic Map